COM448628397 Combi-Blocks, Inc.

(S)-(1-Methyl-azetidin-2-yl)-methanol

  • MDL number MFCD17926451
  • Cas Number : 1310411-24-5
  • Molecular Weight : 101.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9196

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9196
Availability details for
Quantity
  • Availability
  • Add to Cart