COM448628406 Combi-Blocks, Inc.

(S)-1-(5-((S)-2-Carboxypyrrolidin-1-yl)-pentyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD11053583
  • Cas Number : 1820575-05-0
  • Molecular Weight : 298.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9229

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9229
Availability details for
Quantity
  • Availability
  • Add to Cart