COM448628421 Combi-Blocks, Inc.

(S)-1-Benzyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester

  • MDL number MFCD09261309
  • Cas Number : 428518-44-9
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9259

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9259
Availability details for
Quantity
  • Availability
  • Add to Cart