COM448628428 Combi-Blocks, Inc.

(S)-1-Boc-3-n-(pyridin-4-ylmethyl)-amino-piperidine

  • MDL number MFCD07369890
  • Cas Number : 1217722-00-3
  • Molecular Weight : 291.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9272

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9272
Availability details for
Quantity
  • Availability
  • Add to Cart