COM448628433 Combi-Blocks, Inc.

(S)-1-Cbz-3-methyl-1,4-diazepane

  • MDL number MFCD19382354
  • Cas Number : 1311254-86-0
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9279

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9279
Availability details for
Quantity
  • Availability
  • Add to Cart