COM448628434 Combi-Blocks, Inc.

(S)-1-Cyclohexyl-2,2,2-trifluoroethylamine

  • MDL number MFCD08058820
  • Cas Number : 75703-08-1
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9280

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9280
Availability details for
Quantity
  • Availability
  • Add to Cart