COM448628437 Combi-Blocks, Inc.

(S)-1-Isopropyl-piperidine-2-carboxylic acid

  • MDL number MFCD11053660
  • Cas Number : 1141826-51-8
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9285

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9285
Availability details for
Quantity
  • Availability
  • Add to Cart