COM448628438 Combi-Blocks, Inc.

(S)-1-Methyl-2-oxo-imidazolidine-4-carboxylic acid benzyl ester

  • MDL number MFCD08689599
  • Cas Number : 89371-35-7
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9286

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9286
Availability details for
Quantity
  • Availability
  • Add to Cart