COM448628440 Combi-Blocks, Inc.

(S)-1-N-Benzyl-2-cyano-pyrrolidine

  • MDL number MFCD05864626
  • Cas Number : 928056-25-1
  • Molecular Weight : 186.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9288

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9288
Availability details for
Quantity
  • Availability
  • Add to Cart