COM448628443 Combi-Blocks, Inc.

(S)-1-N-Boc-2-cyano-piperidine

  • MDL number MFCD04115365
  • Cas Number : 242459-44-5
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9291

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9291
Availability details for
Quantity
  • Availability
  • Add to Cart