COM448628444 Combi-Blocks, Inc.

(S)-1-N-Boc-3-(2-hydroxyethyl)piperidine

  • MDL number MFCD06796566
  • Cas Number : 863578-32-9
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9292

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9292
Availability details for
Quantity
  • Availability
  • Add to Cart