COM448628447 Combi-Blocks, Inc.

(S)-1-N-Boc-4-N-Fmoc-piperazine-2-carboxylic acid

  • MDL number MFCD02179101
  • Cas Number : 1034574-30-5
  • Molecular Weight : 452.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9295

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9295
Availability details for
Quantity
  • Availability
  • Add to Cart