COM448628449 Combi-Blocks, Inc.

(S)-1-N-Boc-piperidine-2-ethanol

  • MDL number MFCD02683200
  • Cas Number : 199942-74-0
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9297

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9297
Availability details for
Quantity
  • Availability
  • Add to Cart