COM448628451 Combi-Blocks, Inc.

(S)-1-N-Cbz-2-cyano-pyrrolidine

  • MDL number MFCD05864629
  • Cas Number : 63808-36-6
  • Molecular Weight : 230.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9299

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9299
Availability details for
Quantity
  • Availability
  • Add to Cart