COM448628458 Combi-Blocks, Inc.

(S)-1-Pyrrolidin-2-benzyl-2-(n-cbz-n-methyl)amino-ethane

  • MDL number MFCD03426453
  • Cas Number : 675602-74-1
  • Molecular Weight : 352.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9307

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9307
Availability details for
Quantity
  • Availability
  • Add to Cart