COM448628462 Combi-Blocks, Inc.

(S)-1-Pyrrolidin-2-methyl-2-(n-cbz-n-methyl)amino-ethane

  • MDL number MFCD03426451
  • Cas Number : 675602-79-6
  • Molecular Weight : 276.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9311

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9311
Availability details for
Quantity
  • Availability
  • Add to Cart