COM448628468 Combi-Blocks, Inc.

(S)-2-(Aminoethyl)-1-N-Boc-pyrrolidine

  • MDL number MFCD04115291
  • Cas Number : 239483-09-1
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9324
Availability details for
Quantity
  • Availability
  • Add to Cart