COM448628469 Combi-Blocks, Inc.

(S)-2-(Aminomethyl)-1-ethylpyrrolidine hcl

  • MDL number MFCD09991568
  • Molecular Weight : 201.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9325

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9325
Availability details for
Quantity
  • Availability
  • Add to Cart