COM448628471 Combi-Blocks, Inc.

(S)-2,2-Dimethyl-1-trifluoromethyl-propylamine

  • MDL number MFCD08058822
  • Cas Number : 736127-08-5
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9336

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9336
Availability details for
Quantity
  • Availability
  • Add to Cart