COM448628475 Combi-Blocks, Inc.

(S)-2,6-Diamino-hexanoic acid benzyl ester DiHCl

  • MDL number MFCD11501205
  • Cas Number : 16142-09-9
  • Molecular Weight : 309.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9341

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9341
Availability details for
Quantity
  • Availability
  • Add to Cart