COM448628478 Combi-Blocks, Inc.

(S)-2-Amino-3-(3,5-dimethoxy-phenyl)-propionic acid, HCl

  • MDL number MFCD17926412
  • Cas Number : 1391448-08-0
  • Molecular Weight : 261.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9345

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9345
Availability details for
Quantity
  • Availability
  • Add to Cart