COM448628484 Combi-Blocks, Inc.

(S)-2-Benzyl-2-n-bocamino-ethyl thiol

  • MDL number MFCD06796592
  • Cas Number : 141437-85-6
  • Molecular Weight : 267.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9354

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9354
Availability details for
Quantity
  • Availability
  • Add to Cart