COM448628496 Combi-Blocks, Inc.

(S)-2-N-Fmoc-aminomethyl pyrrolidine, HCl

  • MDL number MFCD05861483
  • Cas Number : 1217813-15-4
  • Molecular Weight : 358.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9373
Availability details for
Quantity
  • Availability
  • Add to Cart