COM448628501 Combi-Blocks, Inc.

(S)-3-(2-Chlorophenyl)-beta-alaninol

  • MDL number MFCD07784071
  • Cas Number : 21464-51-7
  • Molecular Weight : 185.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9380

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9380
Availability details for
Quantity
  • Availability
  • Add to Cart