COM448628503 Combi-Blocks, Inc.

(S)-3-(3,4-Dihydroxyphenyl)-n,n-dimethyl-2-methylaminopropionamide hbr

  • MDL number MFCD11707111
  • Cas Number : 1263078-19-8
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9382

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9382
Availability details for
Quantity
  • Availability
  • Add to Cart