COM448628507 Combi-Blocks, Inc.

(S)-3-(3-Fluorophenyl)-beta-alaninol

  • MDL number MFCD07784089
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9386

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9386
Availability details for
Quantity
  • Availability
  • Add to Cart