COM448628509 Combi-Blocks, Inc.

(S)-3-(3-Hydroxyphenyl)-beta-alaninol

  • MDL number MFCD07784103
  • Cas Number : 1213299-59-2
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9389

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9389
Availability details for
Quantity
  • Availability
  • Add to Cart