COM448628510 Combi-Blocks, Inc.

(S)-3-(3-Methoxyphenyl)piperidine

  • MDL number MFCD03839846
  • Cas Number : 88784-37-6
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9390

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9390
Availability details for
Quantity
  • Availability
  • Add to Cart