COM448628512 Combi-Blocks, Inc.

(S)-3-(4-Benzyloxy-phenyl)-2-hydroxy-propionic acid methyl ester

  • MDL number MFCD09261300
  • Cas Number : 481072-37-1
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9394

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9394
Availability details for
Quantity
  • Availability
  • Add to Cart