COM448628516 Combi-Blocks, Inc.

(S)-3-(4-Hydroxyphenyl)-2-hydroxypropionic acid

  • MDL number MFCD01632461
  • Cas Number : 23508-35-2
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9400

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9400
Availability details for
Quantity
  • Availability
  • Add to Cart