COM448628517 Combi-Blocks, Inc.

(S)-3-(4-Methoxyphenyl)-beta-alaninol

  • MDL number MFCD07784113
  • Cas Number : 886061-27-4
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9401

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9401
Availability details for
Quantity
  • Availability
  • Add to Cart