COM448628522 Combi-Blocks, Inc.

(S)-3,3,6,6-Tetramethyltetrahydropyrrolo[1,2-c]oxazol-5(3h)-one

  • MDL number MFCD19382367
  • Cas Number : 156088-45-8
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9415

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9415
Availability details for
Quantity
  • Availability
  • Add to Cart