COM448628527 Combi-Blocks, Inc.

(S)-3-Amino-3-(2-nitro-phenyl)-propionic acid

  • MDL number MFCD04113681
  • Cas Number : 732242-02-3
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9430

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9430
Availability details for
Quantity
  • Availability
  • Add to Cart