COM448628538 Combi-Blocks, Inc.

(S)-3-Amino-3-m-tolyl-propan-1-ol

  • MDL number MFCD07784150
  • Cas Number : 1212834-96-2
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9447

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9447
Availability details for
Quantity
  • Availability
  • Add to Cart