COM448628540 Combi-Blocks, Inc.

(S)-3-Amino-3-p-tolylpropan-1-ol, HCl

  • MDL number MFCD17676124
  • Cas Number : 1391438-22-4
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9450

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9450
Availability details for
Quantity
  • Availability
  • Add to Cart