COM448628551 Combi-Blocks, Inc.

(S)-3-Benzyloxy-2-hydroxy-propionic acid methyl ester

  • MDL number MFCD05863882
  • Cas Number : 127744-28-9
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9463
Availability details for
Quantity
  • Availability
  • Add to Cart