COM448628554 Combi-Blocks, Inc.

(S)-3-Cbz-amino-butylamine

  • MDL number MFCD06804539
  • Cas Number : 168828-15-7
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9468

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9468
Availability details for
Quantity
  • Availability
  • Add to Cart