COM448628566 Combi-Blocks, Inc.

(S)-3-Phenyl-pyrrolidine, HCl

  • MDL number MFCD06796635
  • Cas Number : 1094670-20-8
  • Molecular Weight : 183.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 96
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9484
Availability details for
Quantity
  • Availability
  • Add to Cart