COM448628576 Combi-Blocks, Inc.

(S)-4-Benzyl-3-(2-phenylacetyl)oxazolidin-2-one

  • MDL number MFCD11036321
  • Molecular Weight : 295.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9495

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9495
Availability details for
Quantity
  • Availability
  • Add to Cart