COM448628585 Combi-Blocks, Inc.

(S)-5-Amino-4,5,6,7-tetrahydro-1h-indazole hcl

  • MDL number MFCD11053505
  • Cas Number : 1263078-06-3
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9513

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9513
Availability details for
Quantity
  • Availability
  • Add to Cart