COM448628587 Combi-Blocks, Inc.

(S)-6,6-Dimethyl-morpholine-3-carboxylic acid hcl

  • MDL number MFCD11053601
  • Cas Number : 1313277-24-5
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9520

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9520
Availability details for
Quantity
  • Availability
  • Add to Cart