COM448628594 Combi-Blocks, Inc.

(S)-Amino-(4-fluoro-phenyl)-acetic acid, HCl

  • MDL number MFCD06796581
  • Cas Number : 185994-15-4
  • Molecular Weight : 205.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9531

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9531
Availability details for
Quantity
  • Availability
  • Add to Cart