COM448628597 Combi-Blocks, Inc.

(S)-Aziridine-2-carboxylic acid methyl ester

  • MDL number MFCD07369974
  • Cas Number : 75154-69-7
  • Molecular Weight : 101.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9535

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9535
Availability details for
Quantity
  • Availability
  • Add to Cart