COM448628600 Combi-Blocks, Inc.

(S)-Cyclohexyl-hydroxy-phenyl-acetic acid

  • MDL number MFCD08061195
  • Cas Number : 20585-34-6
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9539

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9539
Availability details for
Quantity
  • Availability
  • Add to Cart