COM448628603 Combi-Blocks, Inc.

(S)-Indoline-2-carboxylic acid ethyl ester, HCl

  • MDL number MFCD09033207
  • Cas Number : 82923-81-7
  • Molecular Weight : 227.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9547

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9547
Availability details for
Quantity
  • Availability
  • Add to Cart