COM448628604 Combi-Blocks, Inc.

(S)-Methyl 1,2,3,4-tetrahydro-quinoline-2-carboxylate hcl

  • MDL number MFCD11053719
  • Molecular Weight : 227.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9549

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9549
Availability details for
Quantity
  • Availability
  • Add to Cart