COM448628609 Combi-Blocks, Inc.

(S)-Methyl-(1-phenyl-2-pyrrolidin-1-yl-ethyl)-amine DiHCl

  • MDL number MFCD08061246
  • Cas Number : 136329-39-0
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9565

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9565
Availability details for
Quantity
  • Availability
  • Add to Cart