COM448628613 Combi-Blocks, Inc.

(S)-N-[[3-(3-Fluorophenyl)-2-oxo-5-oxazolidinyl]methyl]acetamide

  • MDL number MFCD09608092
  • Cas Number : 139071-79-7
  • Molecular Weight : 252.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9574

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9574
Availability details for
Quantity
  • Availability
  • Add to Cart