COM448628614 Combi-Blocks, Inc.

(S)-N-1-Boc-n-4-cbz-2-piperazine carboxylic acid

  • MDL number MFCD02179111
  • Cas Number : 138775-03-8
  • Molecular Weight : 364.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9577

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9577
Availability details for
Quantity
  • Availability
  • Add to Cart