COM448628628 Combi-Blocks, Inc.

(S,S)-1-(3-Methyl-2-methylamino-butyl)-pyrrolidin-3-ol DiHCl

  • MDL number MFCD04115363
  • Molecular Weight : 259.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9603

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9603
Availability details for
Quantity
  • Availability
  • Add to Cart